Order Entry
Denmark
Orders LinkContactUsLinkComponent
D-(-)-Fructose, Millipore®

D-(-)-Fructose, Millipore®

Plastic bottle, 250 g
Catalog # 1.04007.0250
Supplier:   Merck
D-(-)-Fructose, Millipore®
Catalog # 1.04007.0250
Supplier:  Merck
CAS Number:  57-48-7
D-(-)-Fructose, Millipore®
D-(-)-Fructose, Millipore®

Plastic bottle, 250 g

CAS Number:  57-48-7
Price
312,00 kr
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  D(-)-Levulose, (3S,4R,5R)-1,3,4,5,6-pentahydroxyhexan-2-one, β-D-(-)-Fructose, Fructose
  • CAS Number:57-48-7
  • Formula:C₆H₁₂O₆
  • MDL Number:MFCD00148910
  • Molecular Weight:180.16 g/mol
  • Merck Index:13,04295
  • Boiling Pt:552 °C (1013 hPa)
  • Melting Pt:100…110 °C
  • Density:1.59 g/cm³ (20 °C)
  •  :M-Clarity™ quality level = MQ100
  • Storage Temperature:Ambient
  • EINECS:200-333-3

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    250 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.