Estradiol
Glass bottle for solids, 5 g
SERA31100.02
Marketsource
: Serva
: beta-Estradiol, 17β-Estradiol
- :50-28-2
- :C₁₈H₂₄O₂
- :MFCD00003693
- :272.39 g/mol
- :200-023-8
- Pack type:Glass bottle for solids
- Pk:5 g
Anbefalinger