Order Entry
Denmark
Orders LinkContactUsLinkComponent
Estradiol

Estradiol

Glass bottle for solids, 1 g
Catalog # SERA31100.01
Marketsource
Supplier:   Serva
Estradiol
Catalog # SERA31100.01
Supplier:  Serva
Marketsource
CAS Number:  50-28-2
Estradiol

Glass bottle for solids, 1 g

CAS Number:  50-28-2
Price
Spørg for pris
Log ind for tilgængelighed
1 * 1 g
Product Details & Documents
Synonymer:  beta-Estradiol, 17β-Estradiol
  • CAS nummer:50-28-2
  • Formel:C₁₈H₂₄O₂
  • MDL nummer:MFCD00003693
  • Molecular Weight:272.39 g/mol
  • EINECS:200-023-8
Specifikationer

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.