Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Natriumazid

250 g
Catalog # SERA30175.03
Toxic
Environment
Marketsource
Supplier:   Serva
Natriumazid
Catalog # SERA30175.03
Supplier:  Serva
Marketsource
CAS Number:  26628-22-8
Natriumazid

250 g

CAS Number:  26628-22-8
Price
560,07 kr
Log in for availability
1 * 250 g
Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Merck Index:13,08653
  • Melting Pt:275 °C
  • Density:1,85 g/cm³ (20 °C)
  • Storage Temperature:Stue temperatur
  • EINECS:247-852-1
  • UN:1687
  • ADR:6.1,II
Specifications

Specifications

  • Pakn:
    250 g

Specifications