Order Entry
Denmark
Orders LinkContactUsLinkComponent
Sodium azide

Sodium azide

100 g
Catalog # SERA30175.01
Toxic
Environment
Marketsource
Supplier:   Serva
Sodium azide
Catalog # SERA30175.01
Supplier:  Serva
Marketsource
CAS Number:  26628-22-8
Sodium azide

100 g

CAS Number:  26628-22-8
Price
268,83 kr
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Sodium triazide
  • CAS Number:26628-22-8
  • Formula:NaN₃
  • MDL Number:MFCD00003536
  • Molecular Weight:65.01 g/mol
  • Merck Index:13,08653
  • Melting Pt:275 °C
  • Density:1.85 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:247-852-1
  • UN:1687
  • ADR:6.1,II
Specifications

Specifications

  • Pk:
    100 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.