Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
2-Mercaptoethanol

2-Mercaptoethanol

Glass bottle, 500 ml
Catalog # SERA28625.02
Toxic
Corrosive
Environment
Marketsource
Supplier:   Serva
2-Mercaptoethanol
Catalog # SERA28625.02
Supplier:  Serva
Marketsource
CAS Number:  60-24-2
2-Mercaptoethanol
2-Mercaptoethanol

Glass bottle, 500 ml

CAS Number:  60-24-2
Price
477,93 kr
Log in for availability
1 * 500 mL
Product Details & Documents
Synonyms:  2-Hydroxyethyl mercaptan, β-Mercaptoethanol
  • CAS Number:60-24-2
  • Formula:HSCH₂CH₂OH
  • MDL Number:MFCD00004890
  • Molecular Weight:78.14 g/mol
  • Merck Index:13,05894
  • Boiling Pt:157 °C (1013 hPa)
  • Melting Pt:–100 °C
  • Density:1,12 g/cm³ (20 °C)
  • Flash Pt:68 °C
  • Storage Temperature:Stue temperatur
  • EINECS:200-464-6
  • UN:2966
  • ADR:6.1,II
Specifications

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    500 ml

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.