Order Entry
Denmark
Orders LinkContactUsLinkComponent
Crystal violet

Crystal violet

Glass bottle for solids, 25 g
Catalog # SERA27335.01
Corrosive
Health
Irritant
Environment
Marketsource
Supplier:   Serva
Crystal violet
Catalog # SERA27335.01
Supplier:  Serva
Marketsource
CAS Number:  548-62-9
Crystal violet

Glass bottle for solids, 25 g

CAS Number:  548-62-9
Price
238,96 kr
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  Methylrosanilide chloride, Basic violet 3, Viocid, N-(4-(Bis(4-(dimethylamino)phenyl)methylene)cyclohexa-2,5-dien-1-ylidene)-N-methylmethanamonium chloride, Violet crystal, Methyl violet 10B, Dimethyl((di(4-(dimethylamino)phenyl)methylen)-2,5-cyclohexadien-1-ylidene)ammonium chloride, CI 42555, 4-[4,4'-Bis(dimethylamino) benzhydrylidene]cyclohexa-2,5-dien-1-ylidene]dimethylammonium chloride, Hexamethyl violet, Hexamethyl-p-rosaniline chloride, Vianin, Gentian violet
  • CAS Number:548-62-9
  • Formula:C₂₅H₃₀ClN₃
  • MDL Number:MFCD00011750
  • Molecular Weight:407.99 g/mol
  • Merck Index:13,04406
  • Melting Pt:189…194 °C
  • Density:1.19 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:208-953-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications