trans-4-Hydroxy-L(-)-proline
Glass bottle for solids, 100 g
Catalog # SERA25834.03
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
Synonyms: L-4-Hydroxyproline, (S)-(-)-trans-4-Hydroxyproline, L(-)-trans-4-Hydroxyproline, 4-Hydroxy-L-Proline, (2S,4R)-4-Hydroxypyrrolidine-2-carboxylic acid, H-Hyp-OH
- CAS Number:51-35-4
- Formula:C₅H₉NO₃
- MDL Number:MFCD00064320
- Molecular Weight:131.13 g/mol
- Merck Index:13,04866
- Boiling Pt:296 °C (1013 hPa)
- Melting Pt:206 °C
- Storage Temperature:Ambient
- EINECS:200-091-9
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



