Formamide
Glass bottle, 100 mL
Catalog # SERA21710.01
Marketsource
Supplier: Serva
Gå til:
Product Details & Documents
Synonymer: Carbamaldehyde
- CAS nummer:75-12-7
- Formel:HCONH₂
- MDL nummer:MFCD00007941
- Molecular Weight:45.04 g/mol
- Merck Index:13,04261
- Kogende Pt:210 °C (1013 hPa)
- Smeltende Pt:2 °C
- Tæthed:1.1334 g/cm³ (20 °C)
- Flash Pt:79 °C
- Opbevaringstemperatur:Refrigerator
- EINECS:200-842-0
Specifikationer
Specifikationer
- Pack type:Glass bottle
- Pk:100 mL



