Order Entry
Denmark
Orders LinkContactUsLinkComponent
Chloramphenicol

Chloramphenicol

Glass bottle for solids, 100 g
Catalog # SERA16785.02
Sundhed
Marketsource
Supplier:   Serva
Chloramphenicol
Catalog # SERA16785.02
Supplier:  Serva
Marketsource
CAS Number:  56-75-7
Chloramphenicol

Glass bottle for solids, 100 g

CAS Number:  56-75-7
Price
485,40 kr
Log ind for tilgængelighed
1 * 100 g
Product Details & Documents
Synonymer:  Chloromycetin®, Tega-Cetin, (R,R)-Chloramphenicol
  • CAS nummer:56-75-7
  • Formel:Cl₂CHCONHCH(CH₂OH)CH(OH)C₆H₄NO₂
  • MDL nummer:MFCD00078159
  • Molecular Weight:323.13 g/mol
  • Merck Index:13,02087
  • Kogende Pt:645 °C (1013 hPa)
  • Smeltende Pt:150 °C
  • Tæthed:1.49 g/cm³ (20 °C)
  • Opbevaringstemperatur:Ambient
  • EINECS:200-287-4
Specifikationer

Specifikationer

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.