Sodium pyruvate
Glass bottle for solids, 500 g
SERA15220.04
Marketsource
: Serva
: 2-Ketopropionic acid sodium salt, 2-Oxopropionic acid sodium salt, Sodium 2-oxopropanoate, Pyruvic acid sodium salt, α-Ketopropionic acid sodium salt
- :113-24-6
- :CH₃COCOONa
- :MFCD00002586
- :110.04 g/mol
- :220…230 °C
- :1.718 g/cm³ (20 °C)
- :Ambient
- :204-024-4
- Pack type:Glass bottle for solids
- Pk:500 g