Agarose
Glass bottle for solids, 25 g
Catalog # SERA11408.02
Marketsource
Supplier: Serva
Jump to:
Product Details & Documents
- CAS Number:9012-36-6
- MDL Number:MFCD00081294
- Melting Pt:60…90 °C
- Storage Temperature:Ambient
- EINECS:232-731-8
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



