Order Entry
Denmark
Orders LinkContactUsLinkComponent
Test kits, silicate, Spectroquant®, Supelco®
Silicate (silicic acid) test, 0,5 - 500 mg/L Si, 1,1 - 1070 mg/L SiO₂, Molybdosilicate, Prove, Nova 60, Multy, Move, 100 Tests
Catalog # 1.00857.0001
Supplier:   Merck
Test kits, silicate, Spectroquant®, Supelco®
Catalog # 1.00857.0001
Supplier:  Merck
CAS Number:  
Test kits, silicate, Spectroquant®, Supelco®
Test kits, silicate, Spectroquant®, Supelco®
Silicate (silicic acid) test, 0,5 - 500 mg/L Si, 1,1 - 1070 mg/L SiO₂, Molybdosilicate, Prove, Nova 60, Multy, Move, 100 Tests
CAS Number:  
Price
1.400,00 kr
Log in for availability
1 * 1 Each
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60 A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Highly stable, ready to use reagent mixtures in a user friendly design
  • Free selection of appropriate cell format according to the measuring range
  • Long shelf life up to 3 years - all at room temperature
  • Automatic selection of the correct method with help of AutoSelector barcode identification
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Specifications
Specifications
  • Chemical method:
    Molybdosilicate
  • Pk:
    100 Tests
  • Description:
    Silicate (silicic acid) test
  • For unit:
    Prove, Nova 60, Multy, Move
  • Range:
    0,5 - 500 mg/L Si, 1,1 - 1070 mg/L SiO₂
  • Cat. No.:
    1.00857.0001
  • M-Clarity quality level:
    MQ100
Specifications