Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

O-rings for flat flanges, DURAN®

Flat flanges DN 100, Red, 110 mm
  201-0162
O-rings for flat flanges, DURAN®
  201-0162
O-rings for flat flanges, DURAN®
O-rings for flat flanges, DURAN®

Flat flanges DN 100, Red, 110 mm

 
533,00 kr
   
1 * 1 Items
 

Silicone core sheathed with FEP, red.

  • Guarantee excellent joint seal
  • Temperature resistant from −200 to +200 °C
  • Excellent chemical resistance

O-rings for flat flanges with groove.

 
  • For:
    Flat flanges DN 100
  • Ø int.:
    110 mm
  • Colour:
    Red
  • Seq:
    02