Order Entry
Denmark
Orders LinkContactUsLinkComponent
Büchner funnels, Nalgene®

Büchner funnels, Nalgene®

83 ml, 55 mm
Catalog # 221-0100
   Low Manufacturing Impact
Büchner funnels, Nalgene®
Catalog # 221-0100
   Low Manufacturing Impact
Büchner funnels, Nalgene®
Büchner funnels, Nalgene®

83 ml, 55 mm

Price
112,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

PP

  • Robust and lightweight
  • Top and bottom can be separated for easy cleaning
  • Autoclavable

For filter paper or membrane filtration discs.

Specifications
  • Capacity:
    83 ml
  • For filter Ø:
    55 mm
  • Cat. No.:
    221-0100
  • Environmental certification:
    IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
  • Environmentally Preferable:
    Low Manufacturing Impact