Dropping bottles, LDPE, Nalgene®
125 ml, 20-415
Catalog # NALG2416-0125
Supplier: Thermo Fisher Scientific
Dropping bottles, LDPE, Nalgene®
Catalog # NALG2416-0125
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
LDPE bottle, PE dropping pipette, neoprene bulb.
- Suitable for dispensing most acids, bases and alcohols
- Good temperature resistance
- Easy handling
Dispense at a rate you control, be it one drop at a time or in a stream, with LDPE bottles with dropper assembly.
Specifications
- Thread:20-415
- Capacity:125 ml
- Cat. No.:NALG2416-0125
- Environmental certification:IRECS; ISO 14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact