Order Entry
Denmark
Orders LinkContactUsLinkComponent
Dropping bottles, LDPE, Nalgene®
60 ml, 20-415
  NALG2416-0060
Dropping bottles, LDPE, Nalgene®
  NALG2416-0060
 :  
Dropping bottles, LDPE, Nalgene®
Dropping bottles, LDPE, Nalgene®
60 ml, 20-415
LDPE bottle, PE dropping pipette, neoprene bulb.
  • Suitable for dispensing most acids, bases and alcohols
  • Good temperature resistance
  • Easy handling
Dispense at a rate you control, be it one drop at a time or in a stream, with LDPE bottles with dropper assembly.
  • Thread:
    20-415
  • Capacity:
    60 ml