Order Entry
Denmark
Orders LinkContactUsLinkComponent
Blanks for separating funnels, DURAN®
Blanks for separating funnels, DURAN®
500 ml
Catalog # SCOT210134404
Supplier:   DWK Life Sciences
Blanks for separating funnels, DURAN®
Catalog # SCOT210134404
Supplier:  DWK Life Sciences
Blanks for separating funnels, DURAN®
Blanks for separating funnels, DURAN®
500 ml
Price
1.160,00 kr
Log in for availability
1 * 10 Items
Product Details & Documents
Made of DURAN®, borosilicate glass 3.3, clear.
  • For glass manipulators
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Capacity:
    500 ml