Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Funnels, analytical, DURAN®

180 mm, 9 mm, 110 mm
Catalog # 221-1713
Supplier:   DWK Life Sciences
Funnels, analytical, DURAN®
Catalog # 221-1713
Supplier:  DWK Life Sciences
Funnels, analytical, DURAN®
Funnels, analytical, DURAN®

180 mm, 9 mm, 110 mm

Price
515,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

For rapid filtration.

Specifications
  • Funnel O-Ø:
    110 mm
  • Stem O-Ø:
    9 mm
  • Stem length:
    180 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.