Order Entry
Denmark
Orders LinkContactUsLinkComponent
Funnels, analytical, DURAN®

Funnels, analytical, DURAN®

150 mm, 9 mm, 80 mm
Katalog # 221-1712
Leverandør:   DWK Life Sciences
Funnels, analytical, DURAN®
Katalog # 221-1712
Leverandør:  DWK Life Sciences
Funnels, analytical, DURAN®
Funnels, analytical, DURAN®

150 mm, 9 mm, 80 mm

Price
310,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

DURAN®, borosilicate glass 3.3, clear.

  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes

For rapid filtration.

Specifikationer
  • Funnel O-Ø:
    80 mm
  • Stem O-Ø:
    9 mm
  • Stem length:
    150 mm
Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.