Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Raschig rings

Raschig ring, 6 mm, 6 mm
  LENZ05127006
Raschig rings
  LENZ05127006
Raschig rings
Raschig rings

Raschig ring, 6 mm, 6 mm

 
713,00 kr
   
1 * 1 kg
 

Soda lime glass.

  • For packing columns
  • Good chemical resistance
  • Good thermal resistance
 
  • Ext.Ø:
    6 mm
  • Description:
    Raschig ring
  • Height:
    6 mm
  • Seq:
    04
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.