Order Entry
Denmark
Orders LinkContactUsLinkComponent
Large Dewar flasks, type C/CAL
Large Dewar flasks, type C/CAL
Glass refill, type 44, Large Dewar flask, 32 C and 32 CAL
Catalog # 478-0771
Supplier:   KGW
Large Dewar flasks, type C/CAL
Catalog # 478-0771
Supplier:  KGW
Large Dewar flasks, type C/CAL
Large Dewar flasks, type C/CAL
Glass refill, type 44, Large Dewar flask, 32 C and 32 CAL
Price
5.490,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Dewar flasks made of borosilicate glass 3.3 DIN/ISO 3585, for storing and tranporting of LN₂, CO₂ and other cooling liquids. Suitable as insulating flasks.
  • With handle or two grips
  • Insulated lid (incl. grip)
  • The Dewar flasks are vacuum insulated and silvered
  • Large aperture
  • Robust design, with quick release clamp
Type C= metal - blue coated
Type CAL= Stucco aluminium
Temperature range: −200 to +200 °C.
Specifications
  • Description:
    Glass refill, type 44
  • Seq:
    19
  • For:
    Large Dewar flask, 32 C and 32 CAL
  • Cat. No.:
    478-0771
  • Model:
    44