Order Entry
Denmark
Orders LinkContactUsLinkComponent
Large Dewar flasks, type C/CAL
Large Dewar flasks, type C/CAL
Glass refill, type 42, Large Dewar flask, 30 C and 30 CAL
Catalog # 478-0769
Supplier:   KGW
Large Dewar flasks, type C/CAL
Catalog # 478-0769
Supplier:  KGW
Large Dewar flasks, type C/CAL
Large Dewar flasks, type C/CAL
Glass refill, type 42, Large Dewar flask, 30 C and 30 CAL
Price
3.760,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Dewar flasks made of borosilicate glass 3.3 DIN/ISO 3585, for storing and tranporting of LN₂, CO₂ and other cooling liquids. Suitable as insulating flasks.
  • With handle or two grips
  • Insulated lid (incl. grip)
  • The Dewar flasks are vacuum insulated and silvered
  • Large aperture
  • Robust design, with quick release clamp
Type C= metal - blue coated
Type CAL= Stucco aluminium
Temperature range: −200 to +200 °C.
Specifications
  • Description:
    Glass refill, type 42
  • Seq:
    17
  • For:
    Large Dewar flask, 30 C and 30 CAL