Containers, wide mouth, square with thread
300 ml, 67,5×67,5×94 mm, 41 mm, 50 mm
215-3917
: Kautex
PVC, square, transparent or brown transparent colours.
- Wide neck opening
- Easy to fill, easy to clean
- Leakproof
- Suitable for food
- Halal certified
Due to the wide neck opening the containers are easy to fill and to clean. Black closure with PE-foam insert. Brown transparent containers are suitable for light-sensitive media.
- Thread:50 mm
- Capacity:300 ml
- Seq:12
- W×D×H:67,5×67,5×94 mm
- Neck I-Ø:41 mm



