Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Laboratory trays

303×151×81 mm
  216-2110
 :   KARTELL
Laboratory trays
  216-2110
 :  KARTELL
Laboratory trays
Laboratory trays

303×151×81 mm

 
94,10 kr
   
1 * 1 Items
 

PS, white.

  • Robust, impact resistant design
  • Heat resistant up to +70 °C
  • Smooth surfaces for easy cleaning

General purpose trays for a wide variety of laboratory uses. Also suitable for use in the food industry.

 
  • Seq:
    09
  • W×D×H:
    303×151×81 mm