Order Entry
Denmark
Orders LinkContactUsLinkComponent
Cuvettes for Amelung®

Cuvettes for Amelung®

Amelung® cuvettes with metallic mixing balls, 1 ml, 18,65/11,25×22,8 mm
Catalog # KART2631
Supplier:   KARTELL
Cuvettes for Amelung®
Catalog # KART2631
Supplier:  KARTELL
Cuvettes for Amelung®
Cuvettes for Amelung®

Amelung® cuvettes with metallic mixing balls, 1 ml, 18,65/11,25×22,8 mm

Price
457,00 kr
Log in for availability
1 * 1.000 Items
Product Details & Documents

Polystyrene cuvettes designed for the determination of coagulation time.

Specifications
  • Ø×H:
    18,65/11,25×22,8 mm
  • Volume:
    1 ml
  • Description:
    Amelung® cuvettes with metallic mixing balls
  • Seq:
    002
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.