Holders for test tubes
9, 18 mm, 220×55×65,0 mm
Catalog # 212-6895
Supplier: KARTELL
Jump to:
Product Details & Documents
PP, white.
- Good thermal and chemical resistance
- Can be used in water baths
- The tubes contents are always visible
Designed for titrations.
Specifications
- For tube Ø:18 mm
- No. of Holes:9
- W×D×H:220×55×65,0 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Base plate that can hold two racks, 202×156×13,5 mmBase plate that can hold two racks, 202×156×13,5 mm212-6897Log in for availability1 * 1 Items - 90,30 kr
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



