Holders for test tubes
10, 16 mm, 220×55×62,5 mm
212-6710
: KARTELL
PP, white.
- Good thermal and chemical resistance
- Can be used in water baths
- The tubes contents are always visible
Designed for titrations.
- For tube Ø:16 mm
- No. of Holes:10
- W×D×H:220×55×62,5 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



