Order Entry
Denmark
ContactUsLinkComponent
Crystal violet Gurr 'Certistain' for microscopical staining 1 * 100 g
Catalog # 340245L
Supplier:  VWR Chemicals
undefined
Crystal violet Gurr 'Certistain' for microscopical staining 1 * 100 g
Catalog # 340245L
Supplier:  VWR Chemicals

Related Information

Frequently Bought Together