Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Formamide 99.5% for analysis

Glass bottle, 1 L
Catalog # ACRO205820010
Health
Formamide 99.5% for analysis
Catalog # ACRO205820010
CAS Number:  75-12-7
Formamide 99.5% for analysis
Formamide 99.5% for analysis

Glass bottle, 1 L

CAS Number:  75-12-7
Price
627,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:13,04261
  • Boiling Pt:210 °C (760 mmHg)
  • Melting Pt:2…3 °C
  • Density:1.133 g/cm³
  • Flash Pt:175 °C
  • Storage Temperature:Ambient
  • EINECS:200-842-0

 

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Infrared spectrum:
    Conforms
  • Water:
    ≤0.05 % (Coulometric)
  • Sulfated ash:
    ≤0.005 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Copper (Cu):
    ≤1 ppm
  • Iron (Fe):
    ≤1 ppm
  • Zinc (Zn):
    ≤1 ppm
  • Heavy metals (as Pb):
    ≤1 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications