Silver nitrate 99.85% for analysis
Glass bottle for solids, 500 g
Supplier: Thermo Fisher Scientific
Silver nitrate 99.85% for analysis
Catalog # ACRO197685000
Supplier: Thermo Fisher Scientific
CAS Number: 7761-88-8
Jump to:
Product Details & Documents
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Boiling Pt:444 °C (760 mmHg)
- Melting Pt:212 °C
- Density:4,35 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-853-9
- UN:1493
- ADR:5.1,II
- REACH:01-2119513705-43
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Loss on drying:≤0.1 % (1 g, 70°C, 2 h)
- Titration with HCl:≥99.80 %
- Chloride (Cl):≤5 ppm
- Nitrite (NO2):≤10 ppm
- Cadmium (Cd):≤1 ppm
- Calcium (Ca):≤10 ppm
- Chromium (Cr):≤1 ppm
- Copper (Cu):≤1 ppm
- Iron (Fe):≤1 ppm
- Lead (Pb):≤1 ppm
- Manganese (Mn):≤1 ppm
- Nickel (Ni):≤1 ppm
- Zinc (Zn):≤1 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:500 g



