Silver nitrate 99.85% for analysis
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
Silver nitrate 99.85% for analysis
Catalog # ACRO197680050
Supplier: Thermo Fisher Scientific
CAS Number: 7761-88-8
Jump to:
Product Details & Documents
Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt
- CAS Number:7761-88-8
- Formula:AgNO₃
- MDL Number:MFCD00003414
- Molecular Weight:169.87 g/mol
- Merck Index:12,08661
- Boiling Pt:444 °C (760 mmHg)
- Melting Pt:212 °C
- Density:4,35 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-853-9
- UN:1493
- ADR:5.1,II
- REACH:01-2119513705-43
Documents
ACRO19768
SDS
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Loss on drying:≤0.1 % (1 g, 70°C, 2 h)
- Titration with HCl:≥99.80 %
- Chloride (Cl):≤5 ppm
- Nitrite (NO2):≤10 ppm
- Cadmium (Cd):≤1 ppm
- Calcium (Ca):≤10 ppm
- Chromium (Cr):≤1 ppm
- Copper (Cu):≤1 ppm
- Iron (Fe):≤1 ppm
- Lead (Pb):≤1 ppm
- Manganese (Mn):≤1 ppm
- Nickel (Ni):≤1 ppm
- Zinc (Zn):≤1 ppm
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



