n-Hexane 95+%, extra pure
Metal drum, 25 L
Supplier: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
Catalog # ACRO197360250
Supplier: Thermo Fisher Scientific
CAS Number: 110-54-3
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:0.659 g/cm³
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 % (K.F.)
- GC:≥95.0 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.005 %
Specifications
- Pack type:Metal drum
- Pk:25 L



