n-Hexane 95+%, extra pure
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
n-Hexane 95+%, extra pure
Catalog # ACRO197360010
Supplier: Thermo Fisher Scientific
CAS Number: 110-54-3
Jump to:
Product Details & Documents
Synonyms: Hexyl hydride, Hexanen, Hexane
- CAS Number:110-54-3
- Formula:H₃C(CH₂)₄CH₃
- MDL Number:MFCD00009520
- Molecular Weight:86.18 g/mol
- Merck Index:13,04712
- Boiling Pt:69 °C (1013 hPa)
- Melting Pt:–95 °C
- Density:0.659 g/cm³
- Flash Pt:–22 °C
- Storage Temperature:Ambient
- EINECS:203-777-6
- UN:1208
- ADR:3,II
Documents
ACRO19736
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 % (K.F.)
- GC:≥95.0 %
- Color scale:≤20 APHA
- Residue after evaporation:≤0.005 %
Specifications
- Pack type:Glass bottle
- Pk:1 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



