(S)-(-)-Nicotine 99+%
Glass bottle for solids, 100 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: (S)-(-)-3-(N-Methyl-2-pyrrolidinyl)pyridine, (S)-(-)-3-(1-Methyl-2-pyrrolidinyl)pyridine, L(-)-Nicotine, L-Nicotine, Nicotine, (-)-Nicotine
- CAS Number:54-11-5
- Formula:C₁₀H₁₄N₂
- MDL Number:MFCD00006369
- Molecular Weight:162.23 g/mol
- Merck Index:12,06611
- Boiling Pt:247 °C (760 mmHg)
- Melting Pt:–80 °C
- Density:1.015 g/cm³
- Flash Pt:101 °C
- Storage Temperature:Ambient
- EINECS:200-193-3
- UN:1654
- ADR:6.1,II
Specifications
Specification Test Results
- Appearance:Clear colorless to yellow to brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5255 to 1.5285 (20°C, 589 nm)
- Water:≤0.5 % (K.F.)
- GC:≥99.0 %
- Specific optical rotation:-140° to -152° (20°C, 589 nm) (c=2,absolute ethanol) (on anhydrous basis)
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



