Cellulose triacetate
Plastic bottle, 500 g
Appearance: White to light yellow Pellets or rods
- :9012-09-3
- :C₄₀H₅₄O₂₇
- :966.845 g/mol
- :Ambient
- Appearance:White to light yellow Pellets or rods
- Infrared spectrum:Conforms
- Acetyl content:43 to 44 wt %
- Titration Argentometric:≥95 % (on dry substance)
- Water:≤10 %
- Pack type:Plastic bottle
- Pk:500 g



