Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

Methanol 99+%, extra pure

Glass bottle, 2,5 L
Catalog # ACRO177150025
Toxic
Health
Flammable
Methanol 99+%, extra pure
Catalog # ACRO177150025
CAS Number:  67-56-1
Methanol 99+%, extra pure
Methanol 99+%, extra pure

Glass bottle, 2,5 L

CAS Number:  67-56-1
Price
515,00 kr
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Wood alcohol, Methyl alcohol, Hydroxy methane, Carbinol
  • CAS Number:67-56-1
  • Formula:CH₃OH
  • MDL Number:MFCD00004595
  • Molecular Weight:32.04 g/mol
  • Merck Index:13,05984
  • Boiling Pt:64.6 °C (1013 hPa)
  • Melting Pt:–98 °C
  • Density:0.791 g/cm³
  • Flash Pt:12 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-659-6
  • UN:1230
  • ADR:3,II
  • REACH:01-2119433307-44
Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.2 % (K.F.)
  • GC:
    ≥99.0 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.005 %
  • Calcium (Ca):
    ≤0.005 %
  • Copper (Cu):
    ≤0.001 ù
  • Iron (Fe):
    ≤0.002 %
  • Lead (Pb):
    ≤0.005 %
  • Magnesium (Mg):
    ≤0.005 %
  • Manganese (Mn):
    ≤0.0005 %
  • Mercury (Hg):
    ≤0.000001 %
  • Nickel (Ni):
    ≤0.0005 %
  • Potassium (K):
    ≤0.005 %
  • Sodium (Na):
    ≤0.005 %
  • Insoluble matter:
    ≤0.005 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications