Cyclohexane 99.5% for analysis
Glass bottle, 1 L
Supplier: Thermo Fisher Scientific
Cyclohexane 99.5% for analysis
Catalog # 176810010.
Supplier: Thermo Fisher Scientific
CAS Number: 110-82-7
Jump to:
Product Details & Documents
Synonyms: Hexahydrobenzene, Hexamethylene
- CAS Number:110-82-7
- Formula:C₆H₁₂
- MDL Number:MFCD00003814
- Molecular Weight:84.16 g/mol
- Merck Index:13,02752
- Boiling Pt:81 °C (1013 hPa)
- Melting Pt:6.5 °C
- Density:0.77 g/cm³
- Flash Pt:–18 °C
- Storage Temperature:Ambient
- EINECS:203-806-2
- UN:1145
- ADR:3,II
Documents
ACRO17681
SDS
Specifications
Specification Test Results
- Appearance:Clear liquid
- Water:≤0.05 %
- GC:≥99.5 %
- Color scale:≤10 APHA
- Residue after evaporation:≤0.005 %
- Substance darkened by H2SO4:≤10 APHA
- Calcium (Ca):≤0.5 ppm
- Copper (Cu):≤0.1 ppm
- Iron (Fe):≤0.1 ppm
- Lead (Pb):≤0.1 ppm
- Potassium (K):≤0.2 ppm
- Sodium (Na):≤1 ppm
- Zinc (Zn):≤0.1 ppm
Specifications
- Pack type:Glass bottle
- Pk:1 L



