Acrylamide 98.5%, extra pure
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Acrylamide 98.5%, extra pure
Catalog # ACRO164835000
Supplier: Thermo Fisher Scientific
CAS Number: 79-06-1
Jump to:
Product Details & Documents
Synonyms: Prop-2-enamide
- CAS Number:79-06-1
- Formula:C₃H₅NO
- MDL Number:MFCD00008032
- Molecular Weight:71.08 g/mol
- Boiling Pt:125 °C (25 mmHg)
- Melting Pt:82…86 °C
- Density:1.122 g/cm³
- Flash Pt:138 °C
- Storage Temperature:Refrigerator
- EINECS:201-173-7
- UN:2074
- ADR:6.1,III
Specifications
Specification Test Results
- Appearance:White Crystalline powder or crystals
- Infrared spectrum:Conforms
- Melting point:82 to 86 °C
- Water:≤2.0 %
- GC:≥98.0 %
- Color of solution:(10 % in water) Clear and colorless
Specifications
- Pack type:Plastic bottle
- Pk:500 g



