Pyrogallol 99%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Benzene-1,2,3-triol, 1,2,3-Trihydroxybenzene
- CAS nummer:87-66-1
- Formel:C₆H₆O₃
- MDL nummer:MFCD00002192
- Molecular Weight:126.11 g/mol
- Merck Index:13,08090
- Kogende Pt:309 °C (1013 hPa)
- Smeltende Pt:131…135 °C
- Tæthed:1,453 g/cm³ (20 °C)
- Opbevaringstemperatur:Ambient
- EINECS:201-762-9
- FN:2811
- ADR:6.1,III
Specifikationer
Specifikationstestresultater
- Appearance:White to off-white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:131 to 135 °C
- Sulfated ash:≤0.1 %
- HPLC:≥98.5 %
- Arsenic (As):≤0.5 ppm
- Calcium (Ca):≤20 ppm
- Iron (Fe):≤5 ppm
- Magnesium (Mg):≤5 ppm
- Potassium (K):≤20 ppm
- Heavy metals (as Pb):≤5 ppm
- Nitrogen compounds (as N):≤5 ppm
Specifikationer
- Pack type:Plastic bottle
- Pk:500 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



