(±)-Camphorquinone 99%
Glass bottle, 5 g
: 1,7,7-Trimethylbicyclo[2.2.1]heptane-2,3-dione, DL-Camphoroquinone
- :10373-78-1
- :C₁₀H₁₄O₂
- :MFCD00064160
- :166.22 g/mol
- :198…202 °C
- :Refrigerator
- :233-814-1
- Appearance:Yellow Crystalline powder
- Infrared spectrum:Conforms
- Melting point:199 to 204 °C
- GC:≥98.5 %
- Pack type:Glass bottle
- Pk:5 g
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



