Valeric acid 99%
Glass bottle, 5 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Pentanoic acid
- CAS Number:109-52-4
- Formula:C₅H₁₀O₂
- MDL Number:MFCD00004413
- Molecular Weight:102.13 g/mol
- Merck Index:12,10042
- Boiling Pt:185 °C (760 mmHg)
- Melting Pt:20…18 °C
- Density:0.94 g/cm³
- Flash Pt:86 °C
- Storage Temperature:Ambient
- EINECS:203-677-2
- UN:3265
- ADR:8,II
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4070 to 1.4100 (20°C, 589 nm)
- Water:≤0.1 %
- GC:≥98.5 %
Specifications
- Pack type:Glass bottle
- Pk:5 mL
Specifications
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



