Order Entry
Denmark
Orders LinkContactUsLinkComponent
Valeric acid 99%

Valeric acid 99%

Glass bottle, 1 L
Katalog # ACRO149570010
Corrosive
Leverandør:   Thermo Fisher Scientific
Valeric acid 99%
Katalog # ACRO149570010
CAS nummer:  109-52-4
Valeric acid 99%
Valeric acid 99%

Glass bottle, 1 L

CAS nummer:  109-52-4
Price
557,00 kr
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Pentanoic acid
  • CAS Number:109-52-4
  • Formula:C₅H₁₀O₂
  • MDL Number:MFCD00004413
  • Molecular Weight:102.13 g/mol
  • Merck Index:12,10042
  • Boiling Pt:185 °C (760 mmHg)
  • Melting Pt:20…18 °C
  • Density:0.94 g/cm³
  • Flash Pt:86 °C
  • Storage Temperature:Ambient
  • EINECS:203-677-2
  • UN:3265
  • ADR:8,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4070 to 1.4100 (20°C, 589 nm)
  • Water:
    ≤0.1 %
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.