(±)-4-Methyl-2-pentanol 99+%
Glass bottle, 500 mL
Supplier: Thermo Fisher Scientific
(±)-4-Methyl-2-pentanol 99+%
Catalog # ACRO149385000
Supplier: Thermo Fisher Scientific
CAS Number: 108-11-2
Jump to:
Product Details & Documents
Synonyms: (±)-Methyl isobutyl carbinol
- CAS Number:108-11-2
- Formula:C₆H₁₄O
- MDL Number:MFCD00004550
- Molecular Weight:102.18 g/mol
- Boiling Pt:132 °C (760 mmHg)
- Melting Pt:–90 °C
- Density:0.8 g/cm³
- Flash Pt:41 °C
- Storage Temperature:Ambient
- EINECS:203-551-7
- UN:2053
- ADR:3,III
Documents
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4100 to 1.4120 (20°C, 589 nm)
- GC:≥99.0 %
- Specific optical rotation:+28° to +32° (20°C, 589 nm) (c=1 in DMF)
Specifications
- Pack type:Glass bottle
- Pk:500 mL



