Capillary dispensers
Disposable capillary pipettes 5,0 µl, on belts, 5×100 pack
Catalog # 552-0071
Supplier: CAMAG
Jump to:
Product Details & Documents
Compact capillary dispensers can be used on the Nanomat or independently. The capillary pipettes are loaded into the dispenser in magazines from a package of 100 capillaries. Capillaries of 0,5, 1, 2, and 5 µl volume are available in boxes of 500 capillaries (five belts).
- Easy to use
- All components are interchangeable from old to new systems
* With universal capillary holder (552-0044), a dispenser magazine for 1 µl capillaries (552-0065), and one box of 5×100 capillary pipettes of 1,0 µl (552-0069).
Specifications
- Description:Disposable capillary pipettes 5,0 µl, on belts, 5×100 pack
- Seq:11