Order Entry
Denmark
Orders LinkContactUsLinkComponent
Capillary dispensers
Dispenser magazine for 1,0 µl capillaries
  552-0065
 :   CAMAG
Capillary dispensers
  552-0065
 :  CAMAG
Capillary dispensers
Capillary dispensers
Dispenser magazine for 1,0 µl capillaries
2.430,00 kr
1 * 1 Items
Compact capillary dispensers can be used on the Nanomat or independently. The capillary pipettes are loaded into the dispenser in magazines from a package of 100 capillaries. Capillaries of 0,5, 1, 2, and 5 µl volume are available in boxes of 500 capillaries (five belts).
  • Easy to use
  • All components are interchangeable from old to new systems
* With universal capillary holder (552-0044), a dispenser magazine for 1 µl capillaries (552-0065), and one box of 5×100 capillary pipettes of 1,0 µl (552-0069).
  • Description:
    Dispenser magazine for 1,0 µl capillaries
  • Seq:
    05