Order Entry
Denmark
Orders LinkContactUsLinkComponent
Spermidine 99%

Spermidine 99%

Glass bottle for solids, 1 g
Catalog # ACRO132740010
Corrosive
Spermidine 99%
Catalog # ACRO132740010
CAS Number:  124-20-9
Spermidine 99%
Spermidine 99%

Glass bottle for solids, 1 g

CAS Number:  124-20-9
Price
kr387.00
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  N1-(3-Aminopropyl)butane-1,4-diamine, N-(3-Aminopropyl)-1,4-diaminobutane, 4-Azaoctamethylenediamine
  • CAS Number:124-20-9
  • Formula:C₇H₁₉N₃
  • MDL Number:MFCD00008229
  • Molecular Weight:145.25 g/mol
  • Merck Index:12,08893
  • Boiling Pt:128…130 °C (14 mmHg)
  • Melting Pt:22…25 °C
  • Density:0.925 g/cm³
  • Flash Pt:>110 °C
  • Storage Temperature:Refrigerator
  • EINECS:204-689-0
  • UN:3259
  • ADR:8,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear colorless Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4785 to 1.4805 (20°C, 589 nm)
  • GC:
    ≥98.5 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications