Order Entry
Denmark
Orders LinkContactUsLinkComponent
 

D-(-)-Ribose 99+%

Glass bottle for solids, 100 g
Catalog # ACRO132361000
D-(-)-Ribose 99+%
Catalog # ACRO132361000
CAS Number:  50-69-1
D-(-)-Ribose 99+%
D-(-)-Ribose 99+%

Glass bottle for solids, 100 g

CAS Number:  50-69-1
Price
1.830,00 kr
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:50-69-1
  • Formula:C₅H₁₀O₅
  • MDL Number:MFCD00135453
  • Molecular Weight:150.13 g/mol
  • Merck Index:12,08371
  • Boiling Pt:331 °C (1013 hPa)
  • Melting Pt:88…92 °C
  • Density:0,8 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-059-4
Specifications

Specification Test Results

  • Appearance:
    White to light yellow to light beige Crystalline powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤1 % (40°C, 5 h, 20 Torr)
  • Sulfated ash:
    ≤0.1 %
  • HPLC:
    ≥99.0 % (ELSD)
  • Specific optical rotation:
    -19.2° to -20.8° (20°C, 589 nm) (c=4, H2O)
  • Heavy metals (as Pb):
    ≤10 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications

Anbefalinger

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.