3-Methylpentane 99+%
Glass bottle, 250 mL
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
- CAS Number:96-14-0
- Formula:C₆H₁₄
- MDL Number:MFCD00009342
- Molecular Weight:86.18 g/mol
- Boiling Pt:64 °C (760 mmHg)
- Melting Pt:–118 °C
- Density:0.66 g/cm³
- Flash Pt:–32 °C
- Storage Temperature:Ambient
- EINECS:202-481-4
- UN:1208
- ADR:3,II
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3755 to 1.3775 (20°C, 589 nm)
- GC:≥99.0 %
- Assay:≤8 % (Tetracyclin hydrochloride) On dry substance
- Water:≤2.0 %
- Sulfated ash:≤0.5 %
- Identification:Passes test
- pH:2.3 to 3.3 (1 % in water CO2 free)
- Specific optical rotation:-235° to -250° (20°C, 589 nm) (c=0.25, water) (on dry substance)
- Absorbance:≤0.40 (460 nm)
- Heavy metals (as Pb):≤50 ppm
Specifications
- Pack type:Glass bottle
- Pk:250 mL



