Methyl-4-hydroxybenzoate 99%
Plastic bottle, 500 g
Supplier: Thermo Fisher Scientific
Methyl-4-hydroxybenzoate 99%
Catalog # ACRO126965000
Supplier: Thermo Fisher Scientific
CAS Number: 99-76-3
Jump to:
Product Details & Documents
Synonyms: Methylparaben, 4-Hydroxybenzoic acid methyl ester, Methyl Parahydroxybenzoate, Methyl 4-hydroxybenzoate
- CAS Number:99-76-3
- Formula:HOC₆H₄CO₂CH₃
- MDL Number:MFCD00002352
- Molecular Weight:152.15 g/mol
- Merck Index:13,06129
- Boiling Pt:298.6 °C (760 mmHg)
- Melting Pt:125…128 °C
- Density:1,1208 g/cm³ (137 °C)
- Storage Temperature:Ambient
- EINECS:202-785-7
Documents
Specifications
Specification Test Results
- Appearance:White to almost white Crystalline powder
- Infrared spectrum:Conforms
- Melting point:125 to 128 °C
- HPLC:≥98.5 %
- Residue after ignition:≤0.1 %
Specifications
- Pack type:Plastic bottle
- Pk:500 g



